Order Entry
Denmark
ContactUsLinkComponent
 
Health
Celite® 512, medium filter aid
Catalog # ROTH0050.1
Supplier:  Roth Carl
CAS Number:  
Health
Celite® 512, medium filter aid
Catalog # ROTH0050.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    2,5 kg
  • Seq:
    4076

Specifications

About this item

 :
 :60.08 g/mol
 :GHS08
CAS Number:91053-39-3
 :61790-53-2
Formula:SiO₂