0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Denmark
ContactUsLinkComponent
Silica gel
Silica gel, Pore size: 60 Å, Supelco®
Silica gel, Pore size: 60 Å, Supelco®
:
Merck
Silica gel, Pore size: 60 Å, Supelco®
:
Merck
:
63231-67-4
:
7631-86-9
:
SiO₂
:
MFCD00011232
:
2230
:
60.08 g/mol
:
1710
:
M-Clarity™ quality level = MQ100
:
Ambient
Environmentally Preferable:
Frequently Bought Together
TLC and GLP plates, silica gel 60, unmodified layers, glass backed
: Merck
VWR®, Screw Caps for Short Thread Vials ND9
: VWR Collection
VWR®, Crimp Neck Headspace Vials ND20
: VWR Collection
VWR®, Crimp Neck Vials ND11
: VWR Collection
Florisil® 60-100 mesh for chromatography
: VWR Chemicals
VWR®, Short Thread Vials ND9
: VWR Collection
Air valves for safetycaps generation 2.0
: S.C.A.T.
TLC plates, silica gel 60, unmodified and chemically modified layers, aluminium backed
: Merck
VWR® Standard Micro Syringes
: VWR Collection
VWR®, Snap Cap Vials
: VWR Collection
Silica gel 60, irregular 40 - 63 µm for flash chromatography
: VWR Chemicals
Exhaust filters, V3.0
: S.C.A.T.
VWR®, Screw Neck Vials ND13
: VWR Collection
Filter Exhaust M (6 months), housing made of PE-black, filled with activated carbon, connection GL14 for assembly on Safety-Waste- Caps, splash protection cover with note for exchange after 6 months and label field for noting the exchange date. Pac 1 * 1 items
: VWR Collection