Order Entry
Denmark
ContactUsLinkComponent
Potassium thiocyanate 1 mol/l (1 N) in aqueous solution Reag. Ph. Eur. 1071801
Potassium thiocyanate 1 mol/l (1 N) in aqueous solution Reag. Ph. Eur. 1071801
Catalog # 87922.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Potassium thiocyanate 1 mol/l (1 N) in aqueous solution Reag. Ph. Eur. 1071801
Catalog # 87922.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Potassium rhodanide
 :
 :97.18 g/mol
Storage Temperature:Ambient
EINECS:206-370-1
CAS Number:333-20-0
Formula:KSCN
MDL Number:MFCD00011413