Kaliumthiocyanat concentrate for dilution to 1L, 0.1 M KSCN volumetric solution, Fluka™
Leverandør: Honeywell Chemicals
Certifikater
Om denne vare
Synonymer: Potassium rhodanide
CAS nummer:333-20-0
:
Formel:KSCN
MDL nummer:MFCD00011413
Tæthed:1 g/cm³ (25 °C)
:97.18 g/mol
EINECS:206-370-1
:GHS07
Opbevaringstemperatur:Stue temperatur
Specifikationer
- Environmentally Preferable:
Specifikationer
Specifikationstestresultater
- Factor:0.998 - 1.002
Specifikationstestresultater
Frequently Bought Together