Order Entry
Denmark
Orders LinkContactUsLinkComponent
Dibenz[a,j]anthracene

Dibenz[a,j]anthracene

Glass bottle for solids, 100 mg
Catalog # PROOBCR-095
Dibenz[a,j]anthracene
Catalog # PROOBCR-095
CAS Number:  224-41-9
Dibenz[a,j]anthracene

Glass bottle for solids, 100 mg

CAS Number:  224-41-9
Price
7.420,00 kr
Log in for availability
1 * 100 mg
Product Details & Documents
  • CAS Number:224-41-9
  • Formula:C₂₂H₁₄
  • Molecular Weight:278.35 g/mol
  • EINECS:205-928-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg

Specifications