IPTG (isopropyl-β-D-thiogalactopyranoside), Millipore®
Supplier: Merck Millipore (Calbiochem)
Certificates
About this item
CAS Number:367-93-1
:
Formula:C₉H₁₈O₅S
MDL Number:MFCD00063273
:238.31 g/mol
Boiling Pt:402 °C (1013 hPa)
Melting Pt:109.5…110.5 °C
Storage Temperature:Freezer
EINECS:206-703-0
Frequently Bought Together