Order Entry
Denmark
ContactUsLinkComponent
 
IPTG (isopropyl-β-D-thiogalactopyranoside)
Catalog # SHETIB02100
Supplier:  IBI SCIENTIFIC
CAS Number:  
IPTG (isopropyl-β-D-thiogalactopyranoside)
Catalog # SHETIB02100
Supplier:  IBI SCIENTIFIC
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

 :
 :238.31 g/mol
Boiling Pt:402 °C (1013 hPa)
Melting Pt:109.5…110.5 °C
Storage Temperature:Freezer
EINECS:206-703-0
CAS Number:367-93-1
Formula:C₉H₁₈O₅S
MDL Number:MFCD00063273