Naphthalene ≥99.8% (by GC), zone refined: number of passes 23
Catalog # TCIAN0004-1SAMPLE
Supplier: TCI
CAS Number:
Specifications
- Pk:1
- Seq:0014
Specifications
Specification Test Results
- Purity/Analysis Method:>99.8% (GC)
Specification Test Results
About this item
:
:128.17 g/mol
Boiling Pt:218 °C
Melting Pt:81 °C
Flash Pt:79 °C
UN:1334
ADR:4.1,III
CAS Number:91-20-3
Formula:C₁₀H₈
MDL Number:MFCD00001742