Order Entry
Denmark
Orders LinkContactUsLinkComponent
EasySep™ Human B Cell Isolation Kits
EasySep™ Human B Cell Isolation Kits
Marketsource
Leverandør:   Stemcell Technologies
EasySep™ Human B Cell Isolation Kits
Leverandør:  Stemcell Technologies
Marketsource
Certifikater
EasySep™ Human B Cell Isolation Kits
EasySep™ Human B Cell Isolation Kits
3 tilgængelige muligheder nedenfor
Product Details & Documents
Immunomagnetic isolation of untouched human B cells.
  • Fast, easy-to-use and column-free
  • Up to 96% purity with high recovery
  • Isolated cells are untouched
The EasySep™ Human B Cell Isolation Kit is designed to isolate B cells from fresh or previously frozen peripheral blood mononuclear cells or washed leukapheresis samples by immunomagnetic negative selection. The EasySep™ procedure involves labelling unwanted cells with antibody complexes and magnetic particles. The magnetically labelled cells are separated from the untouched, desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube. This product can be used in place of the EasySep™ Human B Cell Enrichment Kit for even faster cell isolations.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Documents
Product Family Options
Produktinformation
  • Description
    Tilgængelighed
    Price
  • EasySep™ human B cell isolation kit, 1×10⁹ cells
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 7.266,00 kr
    7.266,00 kr
    1 * 1 Each
       7.266,00 kr
    Specifikationer:
    Marketsource
    Marketsource
  • EasySep™ human B cell isolation kit, 1×10¹⁰ cells
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each
    Specifikationer:
    Marketsource
    Marketsource
  • EasySep™ kit with RoboSep™ buffer and RoboSep™ filter tips, 1×10⁹ cells
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 7.736,00 kr
    7.736,00 kr
    1 * 1 Each
       7.736,00 kr
    Specifikationer:
    Marketsource
    Marketsource