EasySep™ Human Plasmacytoid DC Isolation Kits
Immunomagnetic negative selection kit.
- Fast, easy-to-use, and column-free
- Up to 97% purity
- Untouched, viable cells
The EasySep™ Human Plasmacytoid DC Isolation Kit is designed to isolate plasmacytoid dendritic cells (pDCs) from human peripheral blood mononuclear cells (PBMCs) or leukapheresis by immunomagnetic negative selection. The EasySep™ procedure involves labelling unwanted cells with antibody complexes and magnetic particles. The magnetically labelled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube.
This product replaces the EasySep™ Human Plasmacytoid DC Enrichment Kit, for even faster cell isolations and is compatible with a greater number of EasySep™ magnets.
: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Frequently Bought Together