EasySep™ Mouse CD4 Positive Selection Kit II
Supplier: STEMCELL Technologies
Certificates
About this item
Immunomagnetic positive selection of CD4+ T cells from mouse splenocytes or other tissues.
- Fast, easy-to-use, and column-free
- Up to 98% purity
- Isolated cells are not fluorochrome-labelled
The EasySep™ Mouse CD4 Positive Selection Kit II is designed to isolate CD4+ cells from single-cell suspensions of splenocytes or other tissues by positive selection. Desired cells are targeted with antibody complexes recognising CD4 and dextran-coated magnetic particles. labelled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off.
This product replaces the EasySep™ Mouse CD4 Positive Selection Kit for even faster cell isolations and does not result in the labelling of isolated cells with PE.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Frequently Bought Together