EasySep™ Mouse CD138 Positive Selection Kits
Supplier: STEMCELL Technologies
Certificates
About this item
Immunomagnetic positive selection of CD138+ cells (e.g. plasma cells) from mouse splenocytes or other tissues.
- Fast, easy-to-use and column-free
- Up to 97% purity with immunised mice
- Useful for the enrichment of plasma cells
- Compatible with hybridoma generation protocols, including electrofusion
The EasySep™ Mouse CD138 Positive Selection Kit is designed to isolate CD138+ cells from mouse splenocytes, lymph nodes and bone marrow by immunomagnetic positive selection. Desired cells are targeted with antibodies and magnetic particles, and separated without columns using an EasySep™ magnet. Unwanted cells are simply poured off, while desired cells remain in the tube. Isolated cells are immediately available for downstream applications such as flow cytometry, cell culture, and hybridoma generation.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Frequently Bought Together