EasySep™ Mouse CD90.1 Positive Selection Kits
Immunomagnetic positive selection of CD90.1+ cells from mouse splenocytes, lymph nodes or from peripheral blood.
- Fast, and easy-to-use
- Up to 97% purity
- No columns required
- Isolated cells are not fluorochrome-labelled
The EasySep™ Mouse CD90.1 Positive Selection Kit is designed to isolate CD90.1+ (Thy1.1+) cells from single suspensions of splenocytes, lymph nodes and peripheral blood by positive selection. Desired cells are targeted with antibody complexes recognising CD90.1 and dextran-coated magnetic particles. labelled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off.
: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Frequently Bought Together