Order Entry
Denmark
ContactUsLinkComponent
EasySep™ Direct Human Naïve B Cell Isolation Kit
Catalog # STMC19264
undefined
EasySep™ Direct Human Naïve B Cell Isolation Kit
Catalog # STMC19264
Specifications
  • Description:
    EasySep™ direct human Naïve B cell isolation kit, 100 ml whole blood processed
  • Environmentally Preferable:
  • Pk:
    1 KIT
Specifications
About this item
Immunomagnetic isolation of naïve B cells directly from whole blood
  • >99,9% red blood cell (RBC) depletion without the need for density gradient centrifugation, sedimentation, or lysis
  • Fast, easy-to-use and column-free
  • Up to 95% purity of isolated cells
  • Isolated cells are untouched
This kit is used to isolate highly purified naïve B cells directly from human whole blood. Isolated cells are unlabelled with antibodies or particles and undergo minimal manipulation.
This kit targets non-naïve B cells for removal with antibodies recognising specific surface markers. Unwanted cells are labelled with antibodies and EasySep™ Direct RapidSpheres™, and separated using an EasySep™ magnet. Desired cells are simply collected into a new tube and are immediately available for downstream applications such as flow cytometry, culture or DNA/RNA extraction.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').