About this item
Immunomagnetic positive selection of human EpCAM+ cells (e.g. mammary epithelial cells).
- Fast and easy-to-use
- Up to 99% purity
- No columns required
The EasySep™ Human EpCAM Positive Selection Kit II is designed to isolate EpCAM+ cells from fresh or previously frozen human mammary epithelial cell preparations by immunomagnetic positive selection. Desired cells are targeted with antibody complexes recognising EpCAM and dextran-coated magnetic particles. The cocktail also contains an antibody to human Fc receptor to minimise nonspecific binding. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off. The EpCAM+ fraction contains luminal cells, luminal-restricted, and bipotential progenitor cells.
This product replaces the EasySep™ Human EpCAM Positive Selection Kit (STMC18356).