Ordreindtastning
Denmark
ContactUsLinkComponent
EasySep™ Mouse CD45 Positive Selection Kit
EasySep™ Mouse CD45 Positive Selection Kit
Katalog # STMC18945
Leverandør:  STEMCELL Technologies
undefined
EasySep™ Mouse CD45 Positive Selection Kit
Katalog # STMC18945
Leverandør:  STEMCELL Technologies
Specifikationer
  • Beskrivelse:
    EasySep™ Mouse CD45 positive selection kit, 2×10⁹ cells
  • Environmentally Preferable:
  • Pakn:
    1 KIT
Specifikationer
Om denne vare
Immunomagnetic positive selection of CD45+ leukocytes from mouse lung, splenocytes, or other tissues.
  • Fast and easy-to-use
  • Up to 98% purity
  • No columns required
  • Isolated cells are not fluorochrome-labelled
The EasySep™ Mouse CD45 Positive Selection Kit is designed to isolate CD45 cells from single-cell suspensions of splenocytes, lung, or other tissues by positive selection. Desired cells are targeted with antibody complexes recognising CD45 and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the ‘Product Restrictions’).