Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Denmark
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Momelotinib
Momelotinib 97%
Momelotinib 97%
474681000.
:
Thermo Fisher Scientific
New Product
:
Momelotinib 97%
474681000.
:
Thermo Fisher Scientific
New Product
:
Pack type:
Glass bottle
Pk:
100 mg
Seq:
2440
Appearance:
white to yellow solid
Infrared spectrum:
Conforms
HPLC:
≥96.0 %
:
:
414.5 g/mol
:
GHS07
:
Ambient
:
1056634-68-4
:
C₂₃H₂₂N₆O₂
:
MFCD16038899