Order Entry
Denmark
ContactUsLinkComponent
AmberChrom™ hydrogen form 50WX8 50-100 mesh
Corrosive
AmberChrom™ hydrogen form 50WX8 50-100 mesh
Certificates
Corrosive
AmberChrom™ hydrogen form 50WX8 50-100 mesh
Specification Test Results
  • Appearance:
    beige to green-beige to light brown beads
  • Total exchange capacity:
    ≥1.7 eq/l (H form)
  • Water retention capacity:
    50 to 56 % (H form)
  • Particle size:
    50 to 100 mesh