Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Cepharanthine 95%

Irritant
Cepharanthine 95%
Certificates

2 Options Available Below

Product Details & Documents
Synonyms:  (15S,31R)-36,54-Dimethoxy-16,32-dimethyl-15,16,17,18,31,32,33,34-octahydro-2,6-dioxa-1(4,5)-[1,3]dioxolo[4,5-g]isoquinolina-3(7,1)-isoquinolina-5(1,3),7(1,4)-dibenzenacyclooctaphane, 6',12'-Dimethoxy-2,2'-dimethyl-6,7-(methylenebis(oxy))oxyacanthan, [14aS-(14aR*,26aS*)]-2,3,13,14,14a,15,26,26a-octahydro-22,30-dimethoxy-1,14-dimethyl- H-4,6:16,19-Dietheno-21,25-metheno-12H-[1,3]dioxolo[4,5-g]pyrido[2′,3′:17,18][1,10]dioxacycloeicosino[2,3,4-ij]isoquinoline, CEP, O-Methylcepharanoline, (14S,27R)-22,33-dimethoxy-13,28-dimethyl-2,5,7,20-tetraoxa-13,28-diazaoctacyclo[25.6.2.216,19.13,10.121,25.04,8.031,35.014,39]nonatriaconta-1(33),3(39),4(8),9,16(38),17,19(37),21,23,25(36),31,34-dodecaene
  • CAS Number:481-49-2
  • Formula:C₃₇H₃₈N₂O₆
  • MDL Number:MFCD00210482
  • Molecular Weight:606.7 g/mol
  • Storage Temperature:Refrigerator
Specifications for Product Family

Specification Test Results

  • Appearance:
    white to light yellow to beige to light orange crystalline powder
  • Infrared spectrum:
    Conforms
  • HPLC:
    ≥94 %
  • Loss on drying:
    ≤1.5 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 1.230,00 kr
    1.230,00 kr
    1 * 250 mg
       1.230,00 kr

  • Glass bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 3.780,00 kr
    3.780,00 kr
    1 * 1 g
       3.780,00 kr