Order Entry
Denmark
ContactUsLinkComponent
Tiapride hydrochloride
Tiapride hydrochloride
New Product
Certificates
Tiapride hydrochloride
New Product

Specification Test Results

  • Appearance:
    white to almost white crystalline powder
  • Infrared spectrum:
    Conforms
  • HPLC:
    ≥98.5 %
  • Solubility:
    (2.5g in 50 ml water) Clear colorless solution
  • Loss on drying:
    ≤0.5 % (1 g, 105°C)

Specification Test Results

Frequently Bought Together