Order Entry
Denmark
ContactUsLinkComponent
Legionella AB Supplement
Legionella AB Supplement
Catalog # LIOF81099
Supplier:  LIOFILCHEM
New Product
CAS Number:  
Legionella AB Supplement
Catalog # LIOF81099
Supplier:  LIOFILCHEM
New Product
CAS Number:  

Specifications

  • Pk:
    10 Vial
  • Description:
    Legionella AB supplement
  • Storage temperature:
    2...8 °C

Specifications

About this item

Legionella AB supplement is a lyophilised selective supplement used in combination with legionella growth supplement for the preparation of legionella BCYE agar base. The complete medium is used for detection and enumeration of legionella spp from clinical specimens and environmental sources including all kinds of water samples.

Certifications: ISO 11731.
Delivery: Each kit contains ten vials of freeze-dried legionella AB supplement, one instruction sheet.