Order Entry
Denmark
ContactUsLinkComponent
Ammonium heptamolybdate:Ammonium metavanadate (4:0.1)% in nitric acid 20% Reag. Ph. Eur. 1056700
Ammonium heptamolybdate:Ammonium metavanadate (4:0.1)% in nitric acid 20% Reag. Ph. Eur. 1056700
Catalog # 87872.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
Ammonium heptamolybdate:Ammonium metavanadate (4:0.1)% in nitric acid 20% Reag. Ph. Eur. 1056700
Catalog # 87872.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
UN:2031
ADR:8,II