Order Entry
Denmark
ContactUsLinkComponent
β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na₂, reduced form)
β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na₂, reduced form)
Catalog # 424237L
Supplier:  VWR Chemicals
CAS Number:  
undefined
β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na₂, reduced form)
Catalog # 424237L
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    0,5 g
  • Seq:
    3035

Specifications

Specification Test Results

  • Assay:
    Min. 95.0 %
  • Water:
    Max. 8.0 %
  • Na (Sodium):
    5.0 8.0 %
  • Molar extinction (260 nm) (10³ l/mol.cm):
    13.9 14.9
  • Molar extinction (340 nm) (10³ l/mol.cm):
    6.1 6.5
  • Proportion A250/A260 (10 mM Tris; pH 10):
    0.79 0.85
  • Proportion A280/A260 (10 mM Tris; pH 10):
    0.21 0.25
  • Proportion A340/A260 (10 mM Tris; pH 10):
    0.42 0.44

Specification Test Results

About this item

 :
 :709.41 g/mol
EINECS:210-123-3
CAS Number:606-68-8
Formula:C₂₁H₂₇N₇Na₂O₁₄P₂
MDL Number:MFCD00036200