Order Entry
Denmark
ContactUsLinkComponent
 
X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside)
Catalog # SHETIB02264
Supplier:  IBI SCIENTIFIC
CAS Number:  
X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside)
Catalog # SHETIB02264
Supplier:  IBI SCIENTIFIC
CAS Number:  

Specifications

  • Pk:
    1
  • Pk Info:
    100 mg

Specifications

About this item

 :
 :408.63 g/mol
Melting Pt:237…239 °C
Storage Temperature:Freezer
EINECS:230-640-8
CAS Number:7240-90-6
Formula:C₁₄H₁₅BrClNO₆
MDL Number:MFCD00005666