Order Entry
Denmark
ContactUsLinkComponent
X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside)
X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside)
  ROCKXGAL-0100
 :  
X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside)
  ROCKXGAL-0100
 :  
  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg
  • Seq:
    2440
 :
 :408.63 g/mol
 :237…239 °C
 :Freezer
 :230-640-8
 :7240-90-6
 :C₁₄H₁₅BrClNO₆
 :MFCD00005666