Order Entry
Denmark
ContactUsLinkComponent
 
 
(3-((3-Cholamidopropyl)dimethylammonio)-2-hydroxy-1-propanesulphonate) ≥98%, white powder
 
 
(3-((3-Cholamidopropyl)dimethylammonio)-2-hydroxy-1-propanesulphonate) ≥98%, white powder

 

  • Identity Test:
    Passes
  • Purity:
    ≥98%
  • FTIR:
    Conforms to Standard
  • pH (10% aq soln) :
    5,0 - 9,0
  • OD350 nm (1% aq soln):
    <0,04

 

Frequently Bought Together