Order Entry
Denmark
ContactUsLinkComponent
N,N-Diethyl-p-phenylenediammonium sulphate, Supelco®
Irritant
N,N-Diethyl-p-phenylenediammonium sulphate, Supelco®
Supplier:  Merck
Certificates
Irritant
N,N-Diethyl-p-phenylenediammonium sulphate, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    500 g/l (20 °C)
  • Melting Point:
    184 - 186 °C
  • Molar Mass:
    262.33 g/mol
  • Bulk Density:
    580 kg/m3
  • Boiling Point:
    274 - 275 °C (1013 hPa)
  • pH:
    2.1 - 2.4 (10 g/l, H2O, 20 °C)
  • Ignition Point:
    540 °C

Specification Test Results

Related Information

Frequently Bought Together