
N,N-Diethyl-p-phenylenediammonium sulphate
Catalog # EHERC12606680
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:262.33 g/mol
Boiling Pt:274…275 °C (1013 hPa)
Melting Pt:184…186 °C
EINECS:228-500-6
:GHS07
Storage Temperature:Ambient
CAS Number:6283-63-2
Formula:C₁₀H₁₈N₂O₄S
MDL Number:MFCD00012993