Oven Sample Processor, 874
Oven sample processor for 6 ml sample vessels
Catalog # METR28740120
Supplier: METROHM
Jump to:
Product Details & Documents
The 874 Oven Sample Processor is used for automatic thermal sample preparation in Karl Fischer titration.
- Oven sample changer: Automated sample preparation for samples that cannot be titrated directly
The oven method is particularly suitable for samples that do not release their water until higher temperatures have been reached, for sparingly soluble samples, or those that react with the KF reagent.
Specifications
- Description:Oven sample processor for 6 ml sample vessels