Order Entry
Denmark
ContactUsLinkComponent
Thermo Scientific™ Capit-All™ Flex Automated Decapper
Thermo Scientific™ Capit-All™ Flex Automated Decapper
Catalog # MATR4120-FLEX
New Product
undefined
Thermo Scientific™ Capit-All™ Flex Automated Decapper
Catalog # MATR4120-FLEX
New Product
Specifications
  • Description:
    Full sevice automated capping and decapping
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
A full-service, automated capping and decapping solution for Thermo Scientific™ Matrix™ and Nunc™ 48 and 96 tube racks.
  • Flexibility: Designed for use as a standalone benchtop unit or in an integrated system
  • Compatible with Matrix and Nunc tubes in 48- and 96-tube formats, can switch between tube formats without adapters, supporting both current and changing application requirements
  • Efficiency: Completes a capping cycle in less than 60 seconds, can handle both full and partial racks of tubes, offering flexibility for varying workflows
  • Angled, 7" touchscreen with user-friendly interface, enables easy program selection with just a few clicks, even with gloves
  • Exceptional sample protection: Includes a drip tray that acts as a barrier between caps and tubes to help prevent cross-contamination and maintain sample integrity
The Capit-All Flex automated decapper delivers the flexibility needed to power genomics, biobanking, precision medicine, and other storage applications while helping reduce your laboratory’s physical footprint and increasing efficiency.
Ordering information: For a selection of compatible tubes, see the Storage tubes, 2D barcoded, Matrix™, Storage tubes, low volume, 2D barcoded, Matrix™, Matrix™ Tri-Coded Tubes, Cryobank vials and Bank-It™ tube system, Nunc™, and Nunc™ Internally-Threaded Universal Tubes and Rack System series.