Phthalocyanine for organic electronics, purified by sublimation
Supplier: TCI
Certificates
About this item
CAS Number:574-93-6
:
Formula:C₃₂H₁₈N₈
MDL Number:MFCD00005085
:514.55 g/mol
Frequently Bought Together
Hydrogen peroxide 35%, J.T.Baker®
Supplier: Avantor
Formamide, EMSURE® for analysis, Supelco®
Supplier: Merck
n-Hexane ≥95%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Chlorobenzene ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Chloroform ≥99.9% (GC, on anhydrous basis) stabilised, PESTINORM® Supra Trace
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
2-Propanol 70% ≥70% (v/v), GPR RECTAPUR®
Supplier: VWR Chemicals
N,N-Dimethylformamide, dehydrated (max. 0.015% H₂O) ≥99.9% for synthesis
Supplier: VWR Chemicals
Isoamyl alcohol, TECHNICAL
Supplier: VWR Chemicals
Chloroform ≥99.8% stabilised, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Water for TOC, Supelco®
Supplier: Merck
2-Propanol ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Petroleum spirit, 60…80 °C
Supplier: Thermo Fisher Scientific