4-(trans-4-Propylcyclohexyl)phenyl trans-4-(trans-4-propylcyclohexyl)cyclohexanecarboxylate
Supplier: TCI
Certificates
About this item
Synonyms: 4-(4-n-Propylcyclohexyl)phenyl-4'-n-propylbicyclohexyl-4-carboxylate, (Trans,trans)-4'-propyl[1,1'-Bicyclohexyl]-4-carboxyllic acid-4-(trans-4-propylcyclohexyl)phenyl ester, 4-(trans-4-Propylcyclohexyl)phenyl trans-4-(trans-4-propylcyclohexyl)cyclohexanecarboxylate
CAS Number:88038-92-0
:
Formula:C₃₁H₄₈O₂
MDL Number:MFCD00673705
Molecular Weight:452.723 g/mol
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together