Nuclear Green™ Photo-Fixable DCS1 *5 mM DMSO Solution*
Catalog # AATB17570
Supplier: AAT BIOQUEST
New Product
CAS Number:
Specifications
- Pk:1 mg
- Description:Nuclear Green™ Photo-fixable DCS1 *5 mM DMSO solution*
Specifications
About this item
Nuclear Green™ Photo-Fixable DCS1 is a unique fluorogenic, DNA-selective, and cell-impermeant dye for analysing DNA content in dead, fixed, or apoptotic cells.
- Stable for 12 months
Quality tested in-house.
Caution: For research use only.