Order Entry
Denmark
ContactUsLinkComponent
Screen Quest™ Fluorimetric Neutrophil Elastase Inhibitor Screening Kit
Screen Quest™ Fluorimetric Neutrophil Elastase Inhibitor Screening Kit
Catalog # AATB11802
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
Screen Quest™ Fluorimetric Neutrophil Elastase Inhibitor Screening Kit
Catalog # AATB11802
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
Specifications
  • Pk:
    100 Tests
Specifications
About this item
Designed for consistent and dependable results. The Screen Quest™ fluorimetric neutrophil elastase inhibitor screening kit offers a fast, sensitive, and high-throughput method for screening and characterising potential inhibitors of NE.
  • Ex/Em=360/470nm
  • Consistent results
  • Six-month shelf life
The amount of fluorescent product is directly proportional to the enzymatic activity of the neutrophil elastase present in the sample.
Delivery: Shipped with blue ice.
Caution: Research use only.