Transfectamine™ 7000 siRNA Transfection Reagents
New Product
Transfectamine™ 7000 siRNA Transfection reagent provides a reliable and effective solution for siRNA-mediated gene knockdown experiments, efficiently delivering small interfering RNA (siRNA) and microRNA (miRNA) into diverse cell types.
- Consistently high transfection efficiency
- Higher viability of transfected cells
- Does not require special medium
For high-throughput siRNA transfection applications, Transfectamine™ 7000 offers a straightforward workflow: the reagent is mixed with siRNA, applied to cells, incubated, and then the extent of gene knockdown is assessed.
: For research use only.
- Cell specific:Eukaryotic
- DNA and siRNA:Yes
- siRNA:Yes
- Storage temperature:Freeze (< –15 °C); Minimise light exposure
Frequently Bought Together