Order Entry
Denmark
ContactUsLinkComponent
ReadiLink™ KLH Conjugation Kit
ReadiLink™ KLH Conjugation Kit
Catalog # AATB5502-10MG
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
ReadiLink™ KLH Conjugation Kit
Catalog # AATB5502-10MG
Supplier:  AAT BIOQUEST
New Product
CAS Number:  

Specifications

  • Pk:
    2
  • No. of reactions:
    2 reactions (bulk)

Specifications

About this item

Designed for consistent and dependable results. This ReadiLink™ KLH conjugation kit is primarily optimised for the simple preparation of hapten-carrier conjugates for immunisation and antibody production.

  • Contains mcKLH formulated in buffers compatible with the carboxyl-reactive carbodiimide reactions and desalt spin columns, which offer exceptional protein recovery by simple centrifugation step
  • Six-month shelf life

The ReadiLink™ KLH conjugation kit is one-step conjugation of a hapten to a carrier protein using the carboxyl-reactive carbodiimide as the crosslinker.

Delivery: Shipped at RT.
Caution: Research use only.