Order Entry
Denmark
ContactUsLinkComponent
ReadiLink™ Rapid mFluor™ Violet 610 Antibody Labelling Kit
ReadiLink™ Rapid mFluor™ Violet 610 Antibody Labelling Kit
Catalog # AATB1116
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
ReadiLink™ Rapid mFluor™ Violet 610 Antibody Labelling Kit
Catalog # AATB1116
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
Specifications
  • Pk:
    2
  • Includes:
    2 labellings
Specifications
About this item
Microscale optimised for labeling 50 µg antibody per reaction. ReadiLink™ Rapid mFluor™ Violet 610 antibody labelling kits provide a quick and convenient method to label antibodies, proteins (<10 kDa), or other amine-containing biomolecules.
  • Robust protocol
  • Consistent results
  • 12-month shelf life
Designed to provide high quality material for downstream fluorescent applications, including flow cytometry and microscopy.
Delivery: Shipped at RT.
Caution: Research use only.