Supelco® Organic Reference Standard, Dextran
Dextran, Analytical standard, for GPC, 50000, 100 mg
Catalog # 00891-100MG
Supplier: Merck
Jump to:
Product Details & Documents
Specifications
Specifications
- Pk:100 mg
- Standards:Analytical standard, for GPC, 50000
- Description:Dextran
- Cat. No.:00891-100MG
- M-Clarity quality level:MQ100
- Formula:[C₆H₁₀O₅]ₙ
- Storage conditions:2...30 °C
- MDL No.:MFCD00130935
- Flash point:Not applicable
- CAS No.:9004-54-0
- Shipping conditions:STD



