Supelco® Organic Reference Standard, Dextran
Supplier: Merck
About this item
Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.
2 Options Available Below
Product Details & Documents
Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 00269-100MG
- 00269-500MG
- M-Clarity quality level:
- MQ100
- MQ100
- CAS No.:
- 9004-54-0
- 9004-54-0
- MDL No.:
- MFCD00130935
- MFCD00130935
- Formula:
- [C₆H₁₀O₅]ₙ
- [C₆H₁₀O₅]ₙ
- Flash point:
- Not applicable
- Not applicable
- Shipping conditions:
- STD
- STD
- Storage conditions:
- 2...30 °C
- 2...30 °C
Specifications
Product Family Options
Product Information
- StandardsPkAvailabilityPrice
- 00269-100MGDextranSpecifications:
More
Specifications
Cat. No.00269-100MGM-Clarity quality levelMQ100CAS No.9004-54-0MDL No.MFCD00130935Formula[C₆H₁₀O₅]ₙFlash pointNot applicableShipping conditionsSTDStorage conditions2...30 °C - 00269-500MGDextranSpecifications:
More
Specifications
Cat. No.00269-500MGM-Clarity quality levelMQ100CAS No.9004-54-0MDL No.MFCD00130935Formula[C₆H₁₀O₅]ₙFlash pointNot applicableShipping conditionsSTDStorage conditions2...30 °C
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



