Supelco® Organic Reference Standard, Octafluoronaphthalene
Catalog # 44080-U
Supplier: Merck
CAS Number:
Specifications
- Pk:1
- Description:Octafluoronaphthalene
- Storage conditions:2...8 °C
- Hazard class description:H225 - H304 - H315 - H336 - H361f - H373 - H411
- Shipping conditions:STD
Specifications
About this item
- GHS signal: GHS02, GHS08, GHS07, GHS09
Caution: P201 - P210 - P273 - P301 + P310 + P331 - P302 + P352 - P308 + P313