Order Entry
Denmark
ContactUsLinkComponent
Supelco® Organic Reference Standard, Trans-2-Tetradecenoyl-L-Carnitine
Supelco® Organic Reference Standard, Trans-2-Tetradecenoyl-L-Carnitine
  40733-10MG
 :  Merck
 :  
Supelco® Organic Reference Standard, Trans-2-Tetradecenoyl-L-Carnitine
  40733-10MG
 :  Merck
 :  

 

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • Description:
    trans-2-Tetradecenoyl-ʟ-carnitine
  • Storage conditions:
    –20 °C
  • Shipping conditions:
    DICE
  • Synonym(s):
    (2R)-3-Carboxy-N,N,N-trimethyl-2-[[(2E)-1-oxo-2-tetradecen-1-yl]oxy]-1-propanaminium inner salt; trans-2-Myristenoyl-ʟ-carnitine; C14:1-Carnitine
  • Molecular weight:
    369,54