Supelco® TraceCERT® Organic Reference Standard, Tricyclazole
Catalog # 39116-10MG
Supplier: Merck
CAS Number:
Specifications
- Pakn:10 mg
- CAS-nr:41814-78-2
- Beskrivelse:Tricyclazole
- Storage conditions:2...8 °C
- Fareklassebeskrivelse:H301 - H412
- MDL No.:MFCD00072466
- M-Clarity quality level:MQ300
- Synonym(s):5-Methyl[1,2,4]triazolo[3,4-b][1,3]benzothiazole
- Molecular weight:189,24
Specifications
About this item
The tricyclazole CRM may also find the following uses: Determination of tricyclazole content in paddy rice using sliver colloid as a substrate and pyridine as an internal standard by surface-enhanced Raman spectroscopy (SERS). To detect residues of tricyclazole and thiram in pear fruit extracts using SERS. Dissipation studies of penoxsulam tricyclazole and profoxydim in rice paddy systems using HPLC-Diode Array Detection (DAD). Determination of tricyclazole residues in rice using liquid chromatography-tandem mass spectrometry (LC-MS/MS) along with the QuEChERS method of sample preparation. Degradation of tricyclazole (TC) in an aqueous solution and real pesticide industry wastewater (RPW) by electron beam (EB) radiation.
Certifications: The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.
Caution: P264 - P270 - P273 - P301 + P310 - P405 - P501