iFluor® 790-Wheat Germ Agglutinin (WGA) Conjugate
Catalog # AATB25563
Supplier: AAT BIOQUEST
New Product
CAS Number:
Specifications
- Pk:1 mg
- Description:iFluor® 790-Wheat germ agglutinin (WGA) conjugate
Specifications
About this item
iFluor® 790 labelled WGA conjugate exhibits a distinct near-infrared fluorescence characteristic, emitting maximally at 812 nm.
- Stable for 12 months
Wheat germ agglutinin (WGA) is a well-studied lectin known for its binding affinity to N-acetyl-D-glucosamine and sialic acid, making it a valuable tool in various biological applications.
Caution: For research use only.