Supelco® Organic Reference Standard, F.A.M.E. Mix RM-3
O7256-1AMP
: Merck
:
- Concentration:Neat
- Pk:1 Ampoul
- Standards:Certified reference material
- Description:F.A.M.E. mix RM-3
- Storage conditions:–20 °C
- Shipping conditions:DICE
- M-Clarity quality level:MQ100
- Synonym(s):AOCS Reference Mixtures; AOCS No. 3 (RM-3); Oil Reference Standard, AOCS No. 3; Oil Reference Standard, AOCS
- Pk Info:Ampule of 100 mg
Myristic acid methyl ester (C14:0) 1,0% Palmitic acid methyl ester (C16:0) 4,0% Stearic acid methyl ester (C18:0) 3,0% Oleic acid methyl ester (C18:1cis-9) 45,0% Linoleic acid methyl ester (C18:2cis-912) 15,0% Linolenic acid methyl ester (C18:3cis-91215) 3,0% Arachidic acid methyl ester (C20:0) 3,0% Behenic acid methyl ester (C22:0) 3,0% Erucic acid methyl ester (C22:1cis-13) 20,0% Lignoceric acid methyl ester (C24:0) 3,0%.
- GHS signal: GHS05