Order Entry
Denmark
ContactUsLinkComponent
Supelco® Organic Reference Standard, AROCLOR 1221 STANDARD
Supelco® Organic Reference Standard, AROCLOR 1221 STANDARD
Catalog # SK1221-1ML
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, AROCLOR 1221 STANDARD
Catalog # SK1221-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Description:
    Aroclor 1221 standard
  • Storage conditions:
    15...25 °C
  • Shipping conditions:
    STD
  • M-Clarity quality level:
    MQ100

Specifications

About this item

  • GHS signal: GHS02, GHS08, GHS07, GHS09