Order Entry
Denmark
ContactUsLinkComponent
 
Supelco® TraceCERT® Organic Reference Standard, Prallethrin
  CRM56578-50MG
 :  Merck
 :  
Supelco® TraceCERT® Organic Reference Standard, Prallethrin
  CRM56578-50MG
 :  Merck
 :  

 

  • Pk:
    50 mg
  • Standards:
    Certified reference material, TraceCERT®
  • CAS No.:
    23031-36-9
  • Description:
    Prallethrin
  • Storage conditions:
    –20 °C
  • Hazard class description:
    H302 - H331 - H410
  • Shipping conditions:
    DICE
  • MDL No.:
    MFCD03792792
  • M-Clarity quality level:
    MQ300
  • Synonym(s):
    (RS)-2-Methyl-4-oxo-3-prop-2-ynylcyclopent-2-enyl (1RS,3RS;1RS,3SR)-2,2-dimethyl-3-(2-methylprop-1-enyl)cyclopropanecarboxylate
  • Molecular weight:
    300,39
  • Flash point:
    139 °C - Closed cup

 

 

 : This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
 : P261 - P264 - P270 - P273 - P301 + P312 - P304 + P340 + P311