Order Entry
Denmark
ContactUsLinkComponent
Supelco® TraceCERT® Organic Reference Standard, Diethofencarb
Supelco® TraceCERT® Organic Reference Standard, Diethofencarb
Catalog # CRM04342-25MG
Supplier:  Merck
CAS Number:  
Supelco® TraceCERT® Organic Reference Standard, Diethofencarb
Catalog # CRM04342-25MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Standards:
    Certified reference material, TraceCERT®
  • CAS No.:
    87130-20-9
  • Description:
    Diethofencarb
  • Storage conditions:
    15...25 °C
  • Hazard class description:
    H319
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00274591
  • M-Clarity quality level:
    MQ300
  • Synonym(s):
    Isopropyl N-(3,4-diethoxyphenyl)carbamate
  • Molecular weight:
    267,32

Specifications

About this item

Diethofencarb may be used as certified reference material for the following:

  • To develop a dispersive liquid-liquid microextraction method based on the solidification of a floating organic droplet (DLLME-SFO) for simultaneous analysis of diethofencarb and pyrimethanil in apple pulp and peel using liquid chromatography with diode-array detection (HPLC-DAD)
  • To investigate the sensitivity profile of Botrytis cinerea field isolates to zoxamide and evaluate the resistance mechanism involved in cross-resistance relationships between benzamides benzimidazoles and N-phenylcarbamates
  • To develop an ultrasound-assisted surfactant-enhanced emulsification microextraction method for the analysis of diethofencarb and pyrimethanil fungicides in water and fruit juice samples using high performance liquid chromatography-diode array detector/electrospray ionisation mass spectrometry (HPLC-DAD/ESI-MS)
  • Preparation of 3D nanotubes assembly from MoS₂ monolayers (3D MoS₂NTs) for rapid and selective screening of diethofencarb
  • Development of a dispersive liquid-liquid microextraction method for the determination of diethofencarb and pyrimethanil in aqueous samples
Caution: P264 - P280 - P305 + P351 + P338 - P337 + P313