Supelco® TraceCERT® Organic Reference Standard, Cyantraniliprole
Catalog # CRM01303-25MG
Supplier: Merck
CAS Number:
Specifications
- Pk:25 mg
- Standards:Certified reference material, TraceCERT®
- CAS No.:736994-63-1
- Description:Cyantraniliprole
- Storage conditions:2…8 °C
- Hazard class description:H410
- Shipping conditions:STD
- MDL No.:MFCD28053506
- M-Clarity quality level:MQ300
- Synonym(s):3-Bromo-1-(3-chloro-2-pyridyl)-4′-cyano-2′-methyl-6′-(methylcarbamoyl)pyrazole-5-carboxanilide
- Molecular weight:473,71
Specifications
About this item
It is intended to be used as a certified reference material (CRM) for calibration in chromatography and other analytical techniques. Cyantraniliprole certified reference material (CRM) may also find following use:
- Study of DNA damage, oxidative stress, and biological toxicity caused in earthworm by cyantraniliprole
- Development of an isotope-labelled internal standard based method using ultra-high-performance liquid chromatography-tandem mass spectrometry for the analysis of chlorantraniliprole and cyantraniliprole in fruits, vegetables, and cereals
- Simultaneous determination of 8 new generation amide insecticides by ultrahigh performance liquid chromatography-tandem mass spectrometry in complex food matrices
- Estimation of five diamide insecticides using carbon nanotube multiplug filter for sample clean-up and ultrahigh-performance liquid chromatography-tandem mass spectrometry in food matrices
- Simultaneous analysis of five diamide insecticides in edible mushrooms by high-performance liquid chromatography coupled to tandem mass spectrometry (HPLC-MS/MS) and a modified QuEChERS procedure for extraction
Caution: P273 - P391 - P501