Supelco® Organic Reference Standard, Decanoyl-ʟ-carnitine
Katalog # 91521-10MG
Supplier: Merck
CAS Number:
Specifications
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:3992-45-8
- Description:Decanoyl-ʟ-carnitine
- Storage conditions:2…8 °C
- Shipping conditions:STD
- M-Clarity quality level:MQ100
- Synonym(s):(2R)-3-Carboxy-N,N,N-trimethyl-2-[(1-oxodecyl)oxy]-1-propanaminium inner salt; ʟ-Carnitine decanoyl ester; C10-Carnitine
- Molecular weight:315,45