Supelco® TraceCERT® Organic Reference Standard, Elemental Impurities according to ICH Q3D inhalation, Standard 4
Catalog # 91496-100ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:In 12% nitric acid
- Pk:100 mL
- Standards:TraceCERT®, applicable for testing acc. to USP<232>, Ph.Eur. Gen. Chapter 5.20
- Description:Elemental Impurities according to ICH Q3D inhalation, Standard 4
- Storage conditions:No temperature limit
- Hazard class description:H290 - H314
- Shipping conditions:STD
- M-Clarity quality level:MQ300
Specifications
About this item
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Caution: P234 - P280 - P303 + P361 + P353 - P304 + P340 + P310 - P305 + P351 + P338 - P363